Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Standard Aerosolv™ Aerosol Can Disposal System: Cartridges and Filters

Catalog No. 19121126 Shop All Justrite Safety Group Products
Change view
Click to view available options
Product Type:
Activated Carbon Cartridge
Color-Changing Carbon Cartridge
Color-Changing Carbon Cartridge with Check Valve
Color-Changing Combination Coalescing/Carbon Filter
Color-Changing Combination Coalescing/Carbon Filter with Check Valve
Combination Coalescing/Carbon Filter
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-121-126 Color-Changing Combination Coalescing/Carbon Filter with Check Valve
19-077307 Activated Carbon Cartridge
19-077306 Combination Coalescing/Carbon Filter
19-121-125 Color-Changing Carbon Cartridge
19-121-127 Color-Changing Carbon Cartridge with Check Valve
19-822-306 Color-Changing Combination Coalescing/Carbon Filter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-121-126 Supplier Justrite™ Supplier No. 28228
Add to Cart
Add to Cart

For use with Serosolv Can Disposal System

Replaxement cartridges and filters

Specifications

Certifications/Compliance EPA, 40 CRF261.23(a)(6)
For Use With (Application) For Aerosolv Aerosol Can Disposal System
Material Carbon
Product Type Color-Changing Combination Coalescing/Carbon Filter with Check Valve
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.