Learn More
Justrite™ Standard Aerosolv™ Aerosol Can Disposal System: Cartridges and Filters
Shop All Justrite Safety Group ProductsDescription
Replacement cartridges and filters.
Specifications
Specifications
| Certifications/Compliance | EPA, 40CRF261.23(a)(6) |
| For Use With (Application) | For aerosolv aerosol can disposal system |
| Material | Carbon |
| Product Type | Color-Changing Combination Coalescing/Carbon Filter |