Learn More
Justrite™ Sure-Grip™ EX™ Safety Cabinets With Handles, For Flammables
Shop All Justrite Safety Group Products

Description
- Sure-Grip EX flush-mounted door handle with two keys for security
- Handle can be padlocked (padlock not included), no interference with traffic
- Exclusive self-latching feature ensures safe closing of cabinet doors without manually engaging door handle
- Double-wall construction provides 1.5 in. (3.8cm) of insulating air space to help prevent spread of flames
- High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
- Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
- Easy-closing, self-latching doors facilitate fire protection of the cabinet
- Self-latch engages critical three-point latching system for protection under fire conditions
- Two-door models hinged left and right with self-closing mechanism ensures proper closing sequence
- Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 165°F (74°C )
- Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
- SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
- Adjustable leveling feet ensure solid positioning and alignment for tight door closure
- Exterior side panel has built-in grounding connector for easy grounding
- Finished inside and out with durable lead-free powder paint
- Meet OSHA and NFPA requirements
- Color: Yellow
Certifications
FM-approved
Compliance
Meet NFPA Code 30 and OSHA regulations; models with self-close doors also comply with the International Fire Code and NFPA1, FIRE CODE™.
Warranty and Services
10 years
Specifications
Specifications
| Product Type | Flammables Safety Cabinet |
| Dimensions (D x W x H) Interior | 14.56 x 39.5 x 60.5 in. |
| Width (Metric) Exterior | 109 cm |
| Width (Metric) Interior | 100.3 cm |
| Depth (Metric) Interior | 37 cm |
| Depth (Metric) Exterior | 45.7 cm |
| No. of Doors | 2 |
| Height (Metric) Exterior | 165.1 cm |
| Height (Metric) Interior | 153.7 cm |
| No. of Shelves | 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.