Learn More
Cayman Chemical JWH 018 N 1 1dImthylPRYl 10mg
SDP
Supplier:  Cayman Chemical 900100010 MG
Encompass_Preferred
Differs from JWH 018 structurally by having a 1,1-dimethylpropyl group, rather than a pentyl chain, extending from the indole group; intended for forensic purposes
Catalog No. 50-289-5818