Learn More
Cayman Chemical JWH 203 4CLOrophenl IsomR 25mg
SDP
Supplier:  Cayman Chemical 1086725 MG
Encompass_Preferred
An internal standard for the quantification of JWH 018 7-hydroxyindole metabolite by GC- or LC-MS
Catalog No. 50-295-1512
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.