Learn More
Cayman Chemical JWH 210 5ethylnphthyl ISME 1mg
SDP
Supplier:  Cayman Chemical 90010401 MG
Encompass_Preferred
An integrin a2B1 inhibitor a2B1 (EC50 130 nM in CHO cells expressing the human receptor) 8-fold selective for integrin a2B1 over integrin a1B1 as well as laminin 332 40 kDa fibronectin 120 kDa fibronectin and vitronectin at 130 nM inhibits the adhesion of platelets in isolated mouse whole blood to collagen I-coated capillaries at a constant shear rate (EC50 6 UM) decreases lavage fluid leukocyte infiltration in a mouse model of platelet-activating factor-induced air pouch inflammation at 10 mg/kg p.o. decreases ear thickness in a mouse model of ovalbumin- and alum-induced mouse model of skin hypersensitivity at 10 mg/kg
Catalog No. 50-289-5693
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.