Learn More
Cayman Chemical RCS4 N 4hydroxypentyl met 1mg
SDP
Supplier:  Cayman Chemical 109361 MG
Encompass_Preferred
2-Arachidonoyl glycerol (2-AG Item No 62160) has been isolated from porcine brain 4614 and has been characterized as the natural endocannabiNId ligand for the CB1 receptor 5306 Incubation of 2-AG with COX-2 and specific PGH2 isomerases in cell cultures and isolated enzyme preparations results in prostaglandin glyceryl ester formation 11045 The biosynthesis of PGH PGD PGE PGF and TXA-2-glyceryl ester compounds have all been documented The 2-glyceryl ester moiety equilibrates rapidly (within minutes) with the more stable 1-glyceryl ester producing a 1090 21-glyceryl ester mixture in typical aqueous media While the stability and metabolism of PGF2a-1-glyceryl ester has been investigated 10555 little is known about its intrinsic biological activity
Catalog No. 50-295-1380
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.