Learn More
Cayman Chemical AM694 NpentnoIc ACD metabo
SDP
Supplier:  Cayman Chemical 14373500 UG
Encompass_Preferred
A fluorescent cationic dye ex/em spectra 502/525 460/650 and 475/590 nm when bound to dsDNA bound to ssDNA or RNA and under acidic conditions respectively ratio of emission at 590 to emission at 525 nm can be used to quantify the increase in acidic vesicular organelles observed during cellular autophagy
Catalog No. 50-297-0045
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.