Learn More
Enzo Life Sciences JZL-184 (10mg). CAS: 1101854-58-3
Supplier: Enzo Life Sciences BMLEI3910010
Potent, selective and irreversible inhibitor of MAG lipase (IC50=8nM vs 4µM for FAAH). JZL-184-treated mice displayed a number of CB1-dependent behavioral effects including analgesia, hypothermia and hypomotility as well as increased levels of 2-arachidonoylglycerol in the brain ( approx. 8-fold, with no effect on anandamide levels). JZL-184 produces a rapid and sustained blockade of brain 2-AG hydrolase activity in mice. It blocks neuropathic pain and enhances retrograde endocannabinoid signaling. Alternative name: 4-Nitrophenyl-4-(dibenzo[d][1,3]dioxol-5-yl(hydroxy)methyl)piperidine-1-carboxylate. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: Ambient.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.