Learn More
Selleck Chemical LLC JZL184 10mg 1101854-58-3
SDP
Supplier:  Selleck Chemical LLC S490410MG
Encompass_Preferred
JZL 184 is the first selective inhibitor of monoacylglycerol lipase (MAGL) with IC50 of 8 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4295
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.