Learn More
Enzo Life Sciences K-252a (1mg). CAS: 99533-80-9
SDP
Supplier:  Enzo Life Sciences BMLEI1521000
Encompass_Preferred
Alkaloid isolated from soil fungi. General, cell permeable protein kinase inhibitor. Acts by binding to the ATP binding domain of the kinase. Potent inhibitor of Ca2+/calmodulin kinase II. Inhibits myosin light chain kinase, cAMP-dependent protein kinase (PKA), protein kinase C (PKC) (Ki=25nM), trk tyrosine kinase family members (gp140trk; IC50 =3nM) and cGMP-dependent protein kinase (PKG). Binds with high affinity to neuroblastoma membrane (2.7nM), promoting survival of primary neuronal cultures (75nM) and induction of neurite outgrowth in SH-SY5Y neuroblastoma cells along with phosphorylation of pp125 focal adhesion protein tyrosine kinase. Induces apoptosis. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (1mg/ml) or dimethyl formamide (1mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0572
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.