Learn More
AdipoGen K-252c
SDP
Supplier:  AdipoGen AGCN20097M001
Encompass_Preferred
Chemical. CAS 85753-43-1. Formula C20H13N3O. MW 311.4. Isolated from Streptomyces longisporoflavus. Indolocarbazole alkaloid antibiotic. Potent cell permeable reversible and ATP-competitive PKC protein kinase C inhibitor with 10-fold selectivity over PKA protein kinase A. Anticancer compound. Cytotoxic againt various cancer cell lines. Apoptosis inducer. Antiviral compound against GCV-sensitive and -resistant strains of human cytomegalovirus HCMV. LACTB beta-lactamase, malate dehydrogenase MDH2, MDHC, MDH1B, ME1. ME2, ME3 and chymotrypsin inhibitor. Inhibits mixed lineage kinase MLK.
Catalog No. 50-256-2663
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.