Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ K-Ras (G12D Mutant) Recombinant Rabbit Monoclonal Antibody (HL2640)
GREENER_CHOICE

Catalog No. MA556855
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA556855 Supplier Invitrogen™ Supplier No. MA556855
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

K-Ras (G12D Mutant) Recombinant Monoclonal Antibody for ICC/IF, Western Blot

The K-Ras gene, specifically the G12D mutation, is a well-known oncogene implicated in various types of cancer, including pancreatic, colorectal, and lung cancers. The G12D mutation involves a substitution of glycine (G) with aspartic acid (D) at the 12th position of the K-Ras protein. This mutation leads to a constitutively active K-Ras protein that continuously signals for cell growth and division, bypassing normal regulatory mechanisms. As a result, cells harboring this mutation proliferate uncontrollably, contributing to tumorigenesis. The K-Ras (G12D) mutant is a critical target for cancer research and drug development, as targeting this mutation could potentially inhibit the aberrant signaling pathways and slow down or stop cancer progression. Despite the challenges in targeting mutant K-Ras proteins due to their high affinity for GTP/GDP and lack of suitable binding pockets, ongoing research is focused on developing specific inhibitors and novel therapeutic strategies to effectively target this mutation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen K-Ras (G12D Mutant)
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2640
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene KRAS
Gene Accession No. P01116
Gene Alias K RAS; KRAS; K-Ras
Gene Symbols KRAS
Host Species Rabbit
Immunogen Synthetic peptide of human RAS (G12D Mutant).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3845
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.