Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ K-Ras Monoclonal Antibody (F132-62)
GREENER_CHOICE

Catalog No. PIMA191046
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA191046 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA191046 Supplier Invitrogen™ Supplier No. MA191046
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: Sw480 or Y1 cells or normal skin tissue..

K-RAS (GTPase KRas) belongs to the Ras oncogene family. Ras proteins bind GDP/GTP and possess intrinsic GTPase activity. It plays an important role in the regulation of cell proliferation and has a role in promoting oncogenic events by inducing transcriptional silencing of tumor suppressor genes in colorectal cancer cells in a ZNF304-dependent manner. Mutations in the gene can result in leukemia acute myelogenous, leukemia juvenile myelomonocytic, Noonan Syndrome, gastric cancer, and cardiofaciocutaneous syndrome 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen K-Ras
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone F132-62
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation 0.05M sodium phosphate with 0.2% gelatin and 0.1% sodium azide
Gene KRAS
Gene Accession No. P01116, P08644, P32883
Gene Alias AI929937; cellular c-Ki-ras2; cellular c-Ki-ras2 proto-oncogene; CFC2; c-Ki-ras; c-Kirsten-ras protein; c-Kirsten-ras proto-oncogene; c-K-ras; c-K-ras2 protein; fa04e08; fc14b12; fc23g10; fj89d12; GTPase KRas; GTPase KRas, N-terminally processed; hypothetical protein LOC445289; K RAS; ki-Ras; Kirsten rat sarcoma oncogene 2, expressed; Kirsten rat sarcoma viral (Kras-2) oncogene homolog; Kirsten rat sarcoma viral oncogene; Kirsten rat sarcoma viral oncogene homolog; Kirsten rat sarcoma viral oncogene homologue 2 (active); KRAS; K-ras; K-Ras 2; K-ras p21 protein; KRAS proto-oncogene, GTPase; KRAS1; Kras2; Kras-2; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; NS; NS3; oncogene KRAS2; p21; p21 protein; p21B; p21ras; PR310 c-K-ras oncogene; RALD; ras; ras p21; RASK; RASK2; ras-like protein; transforming protein p21; Unknown; v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog; v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog; wu:fa04e08; wu:fc14b12; wu:fc23g10; wu:fj89d12; xRAS2; zgc:85725
Gene Symbols KRAS
Host Species Mouse
Immunogen Recombinant full length protein (Human).
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16653, 24525, 3845
Target Species Human, Mouse, Rat
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.