Learn More
Kawasumi™ K-Shield™ Advantage Small Vein Infusion Sets
Catalog No. 22010182
Change view
Click to view available options
Needle Gauge:
21 G
23 G
25 G
Color:
Dark Blue
Green
Light Blue
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Color
22-010-182 21 G Green
22-010-183 23 G Light Blue
22-010-184 25 G Dark Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-010-182 Supplier Kawasumi™ Supplier No. D3KA21G
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ensures safe collection environment for clinician and patient
  • One-step activation with audible click
  • Needle covered during removal
  • Extra steps for disposal eliminated through integrated safety device
Specifications
Product Type Small Vein Infusion Set
Color Green
Needle Gauge 21 G