Learn More
Selleck Chemical LLC K03861 5mg 853299-07-7 AUZ 454AUZ454
SDP
Supplier:  Selleck Chemical LLC S81005MG
Encompass_Preferred
K03861 (AUZ 454,AUZ454) is a type II CDK2 inhibitor with Kd of 50 nM, 18.6 nM, 15.4 nM, and 9.7 nM for CDK2(WT), CDK2(C118L), CDK2(A144C), and CDK2(C118L/A144C), respectlvely. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5651