Learn More
MOLECULAR DEPOT LLC 2-OH-a-cyclodextrin - 25 mL
Supplier: MOLECULAR DEPOT LLC K1010005B
(2-hydroxypropyl)-alpha-cyclodextrin Solution 10perc (w/v) is an aqueous solution of the cyclic hydroxylated dextrin of type Alpha This product has been used as versatile tools in molecular assemblies enhancement of solubility and organic synthesis It has also been used in a wide array of other biochemical and immunological applications For in vitro research use only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.