Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega K562 Genomic DNA

Catalog No. PRE4931 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
80 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE4931 80 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRE4931 Supplier Promega Supplier No. E4931
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Purified from the human lymphoma cell line K562 by detergent lysis and proteinase K digestion. Supplied at a concentration of 400microg/mL in TE buffer (10mM Tris-HCl, 1mM EDTA; final pH 8.0).

K562 Genomic DNA is supplied at a concentration of 400μg/ml in TE buffer (10mM Tris-HCl, 1mM EDTA; final pH 8.0).

Specifications

Content And Storage 2°C to 10°C
Concentration 400 μg/mL
Quantity 80 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.