Learn More
AdipoGen K777 [K11777] (1 mg)
Supplier: AdipoGen AGCR10158M001
K777 [K11777] (1 mg), AdipoGen Life Sciences. Chemical. CAS: 233277-99-1. Formula: C32H38N4O4S. MW: 574.7. Synthetic. Broad-range cathepsin inhibitor (for all known cathepsins) useful for inflammasome inhibition. Inhibits cathepsins B, L, S, C, V, K and papain in cell-free assays. Inhibits cathepsin X as well. Suppresses particle-induced NLRP3 activation and IL-1beta secretion more efficiently than Ca074Me (cathepsin B inhibitor), which in contrast suppresses nigericin-induced IL-1beta secretion. Selectively inhibits particle-induced cell death but not nigericin- or sAdT-induced cell death. Potent and selective CCR4 antagonist. Potent, irreversible cysteine protease inhibitor. Potent cysteine protease cruzain (cruzipain) inhibitor.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.