Learn More
Cayman Chemical (−)-(α)-Kainic Acid (hydrate), 58002-62-3, 25mg
SDP
Supplier:  Cayman Chemical 7805025
Encompass_Preferred
Molecular Formula C10H15NO4 • H2O, Purity ≥98%, Formula Weight 231.2, Synonyms: 2S-carboxy-4S-(1-methylethenyl)-3S-pyrrolidineacetic acid, monohydrate; A natural marine product originally isolated from the red marine alga D. simplex that is a potent CNS stimulant; developed as the prototype neuroexcitatory amino acid for the induction of seizures in experimental animals, at a typical dose of 10-30 mg/kg in mice; neuroexcitotoxic and epileptogenic, acting through specific kainate receptors.
Catalog No. NC0363501
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.