Learn More
Tocris Bioscience™ Kainic acid
Description
Related compounds include synthetic Kainic acid (Cat. No. 7065), the kainate receptor agonist Domoic acid (Cat. No. 0269) and the antagonist NBQX (Cat. No. 0373).
Specifications
Specifications
| Chemical Name or Material | (2 S,3 S,4 S)-Carboxy-4-(1-methylethenyl)-3-pyrrolidineacetic acid |
| CAS | 487-79-6 |
| Assay Percent Range | >98% |
| Quantity | 1 mg |
| Solubility | Soluble to 25mM in water with gentle warming |
| Target | Kainate Receptor Agonists |
| Molecular Formula | C10H15NO4 |
| Formula Weight | Observed MW: 213.23 |
| Purity | 98% |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.