Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109887 Supplier Invitrogen™ Supplier No. PA5109887
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDPEAYGSPC ARGSQPWQVS LFNGLSFHCA GVLVDQSWVL TAAHCGNKPL WARVGDDHLL LLQGEQLRRT TRSVVHPKYH QGSGPILPRR TDEHDLMLLK LARPVVLGPR VRALQLPYRC AQPGDQCQVA GWGTTAARRV KYNKGLTCSS ITILSPKECE VFYPGVVTNN MICAGLDRGQ DPCQSDSGGP LVCDETLQGI LSWGVYPCGS AQHPAVYTQI CKYMSWINKV IRSN.

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Its encoded protein is secreted and may play a role in suppression of tumorigenesis in breast and prostate cancers. Alternate splicing of this gene results in multiple transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 10
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 6.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KLK10
Gene Accession No. O43240, P36375
Gene Alias 2300002A13Rik; breast normal epithelial cell associated serine protease; kallikrein 10; kallikrein 10 protein 1; kallikrein 10 protein 12; kallikrein 10 protein 13; kallikrein 10 protein 2; kallikrein 10 protein 3; kallikrein 10 protein 4; kallikrein 10 protein 5; kallikrein 10 protein 7; kallikrein 10 protein 8; kallikrein 10 protein 9; kallikrein related peptidase 10; kallikrein related-peptidase 10; kallikrein-10; kallikrein-related peptidase 10; Klk10; NES1; Normal epithelial cell-specific 1; protease serine-like 1; protease, serine-like, 1; PRSSL1
Gene Symbols KLK10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 43-276 of human KLK10 (NP_6658951).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292850, 5655, 69540
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.