Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 12 Antibody (364932) [DyLight 550], R&D Systems™
SDP

Catalog No. FAB40341L
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB40341L 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB40341L Supplier R&D Systems Supplier No. FAB40341L

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Kallikrein 12 Monoclonal antibody specifically detects Kallikrein 12 in Human samples. It is validated for Immunoprecipitation,Western Blot,Neutralization

Specifications

Antigen Kallikrein 12
Applications Immunoprecipitation, Western Blot, Neutralization
Classification Monoclonal
Clone 364932
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias DKFZp686H1078, EC 3.4.21, EC 3.4.21.-, kallikrein 12, Kallikrein-like protein 5, kallikrein-related peptidase 12, KLKL5kallikrein-12, KLK-L5MGC42603
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Kallikrein 12, Ala18-Asn248, Accession # Q9UKR0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 43849
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.