Learn More
Invitrogen™ Kallikrein 2 Monoclonal Antibody (6B7)
Description
This antibody detects Kallikrein 2 in human samples and has no detectable reactivity with PSA.
Specifications
Specifications
| Antigen | Kallikrein 2 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 6B7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Gene | KLK2 |
| Gene Accession No. | P20151 |
| Gene Alias | glandular kallikrein 2; glandular kallikrein-1; hGK-1; hK2; kallikrein 2, prostatic; kallikrein related peptidase 2; kallikrein-2; kallikrein-related peptidase 2; KLK2; KLK2A2; MGC12201; Tissue kallikrein 2; Tissue kallikrein-2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.