Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596350
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596350 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596350 Supplier Invitrogen™ Supplier No. PA596350
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IIDGAPCARG SHPWQVALLS GNQLHCGGVL VNERWVLTAA HCKMNEYTVH LGSDTLGDRR AQRIKASKSF RHPGYSTQTH VNDLMLVKLN SQARLSSMVK KVRLPSRCEP PGTTCTVSGW GTTTSPDVTF PSDLMCVDVK LISPQDCTKV YKDLLENSML CAGIPDSKKN ACNGDSGGPL VCRGTLQGLV SWGTFPCGQP NDPGVYTQVC KFTKWINDTM KKHR; Positive Samples: 293T, HeLa, HepG2, Mouse liver, Mouse heart, Mouse brain, Mouse kidney.

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Its encoded enzyme is thought to be involved in the proteolysis of intercellular cohesive structures preceding desquamation, which is the shedding of the outermost layer of the epidermis. Alternative splicing of this gene results in two transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KLK7
Gene Accession No. P49862, Q91VE3
Gene Alias hK7; hSCCE; kallikrein 7 (chymotryptic, stratum corneum); kallikrein related peptidase 7; kallikrein related-peptidase 7 (chymotryptic, stratum corneum); kallikrein-7; kallikrein-related peptidase 7; KLK7; protease, serine, 6; Prss6; SCCE; Serine protease 6; signal protein; stratum corneum chymotryptic enzyme; Thymopsin
Gene Symbols KLK7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-253 of human KLK7 (NP_6448061).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23993, 5650
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.