Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Kallikrein 8/Neuropsin Recombinant Protein Antigen
SDP

Catalog No. NB398167 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398167 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398167 Supplier Novus Biologicals™ Supplier No. NBP305517PEP
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1 M Urea, pH 7.4.
Gene ID (Entrez) 11202
Molecular Weight (g/mol) MolecularWeight-theroretical: 26 kDa
Quantity 100 μL
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Gene Alias EC 3.4.21, EC 3.4.21.118, hK8, HNP, kallikrein 8 (neuropsin/ovasin), kallikrein-related peptidase 8, KLK8, KLK8 protein type 1, KLK8 protein type 2, Neuropsin, neuropsin type 1, neuropsin type 2, NP, NRPN, Ovasin, PRSS19kallikrein-8, Serine protease 19, Serine protease TADG-14, TADG14neuropsin, Tumor-associated differentially expressed gene 14 protein
Gene Symbol KLK8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Inhibitor Properties This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP3-05517. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL
Form Purified
Research Category Neuroscience
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein Kallikrein 8/Neuropsin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.