Learn More
Bachem H-D-Phe-Phe-Arg-chloromethylketone
SDP
Supplier:  Bachem 4014271.0025
Encompass_Preferred
H-D-Phe-Phe-Arg-chloromethylketone Trifluoroacetate, 74392-49-7, C25H33ClN6O3, Mr 501.03, 25mg. DPhe-Phe-Arg-chloromethylketone. Kallikrein inhibitor which is even effective in preventing apolipoprotein B-100 proteolysis in dextran sulfate (DS)-precipitated human plasma low density lipoproteins (LDL). D-FFR-CMK also inhibits gingipain R.
Catalog No. 50-194-6783
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.