Learn More
Enzo Life Sciences Kanamycin B . sulfate (high purity) (250mg). CAS: 29701-07-3
SDP
Supplier:  Enzo Life Sciences ALX380095M250
Encompass_Preferred
Aminoglycoside antibiotic. Binds to the 70S ribosomal subunit. Inhibits translocation and elicits miscoding. Effective against Gram-negative and Gram-positive bacteria and mycoplasma. Alternative name: Bekanamycin . sulfate. Purity: ≥99% (HPLC). Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8952