Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Kanamycin disulfate, 700μg/mg

Click to view available options
Quantity:
5g
25g
64013-70-3
C18H40N4O19S2
680.647
MFCD00143916
OGTKIXVMLDAMNU-KNQICTBBSA-N
Kanamycin A; Kanamycin acid sulfate
24721551
(2R,3S,4S,5R,6R)-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O.OS(=O)(=O)O
64013-70-3
C18H40N4O19S2
680.647
MFCD00143916
OGTKIXVMLDAMNU-KNQICTBBSA-N
Kanamycin A; Kanamycin acid sulfate
24721551
(2R,3S,4S,5R,6R)-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O.OS(=O)(=O)O
Solubility Information Soluble in aqueous solutions
Formula Weight 680.65
Physical Form Powder
Color Colorless
Melting Point >260°C
Quantity 5g
Sensitivity Moisture & Light Sensitive
Chemical Name or Material Kanamycin disulfate, 700μg/mg

storageNote1 : 2° to 8°C