Learn More
Research Products International Corp Kanamycin Sulfate Sol 5 ml
SDP
Supplier:  Research Products International Corp ZA10035
Encompass_Preferred
Kanamycin inhibits bacterial protein synthesis by binding 70S ribosomes resulting in dysfunctional translation of mRNA. Kanamycin is effective against both Gram (-) and Gram (+) bacteria. Resistance to kanamycin is conferred by an aminoglycoside phosphotransferase that modifies the antibiotic, preventing its interaction with ribosomes. Kanamycin is commonly used to select for cosmid vectors.
Catalog No. 50-238-8464
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.