Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KANK2/SIP Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501560308
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0308 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0308 Supplier Bethyl Laboratories, Inc Supplier No. A301407AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

Ankyrins are membrane adaptor molecules that play important roles in the control of cytoskeleton formation by regulating actin polymerization. KANK2 (KN motif and ankyrin repeat domain-containing protein 2), was initially identified as a steroid receptor coactivator (SRC) interacting protein (SIP) that could sequester steroid receptor coactivators in the cytoplasm. More recent experiments have shown that while KANK2 is widely expressed, in kidney podocytes, KANK2 localizes to foot processes, suggesting that KANK2 may contribute to controlling actin dynamics in podocyte foot processes.

Specifications

Antigen KANK2/SIP
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene KANK2
Gene Accession No. Q63ZY3
Gene Alias KANK2, KN motif and ankyrin repeat domains 2, SRC-interacting protein, SIP, matrix-remodeling-associated protein 3, MXRA3, Ankyrin repeat domain-containing protein 25, ANKRD25
Gene Symbols KANK2
Host Species Rabbit
Immunogen Between 550 and 600
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25959
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.