Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KANK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562333
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562333 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562333 Supplier Invitrogen™ Supplier No. PA562333
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLREATTQTP WSCAEKAAQT ESPAEAPSLT QESSPGSMDG DRAVAPAGIL KSIMK Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 51%.

Ankyrins are membrane adaptor molecules that play important roles in the control of cytoskeleton formation by regulating actin polymerization. Like other members of the KANK family, KANK3 (KN motif and ankyrin repeat domain-containing protein 3), is thought to play a role in the formation of actin stress fibers. In zebrafish, the homolog of KANK3 interacts with the adaptor protein Numb, a protein implicated in multiple basic cellular processes, and is essential for epidermal integrity and neurulation, suggesting that KANK3 may play a similar role in higher organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KANK3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KANK3
Gene Accession No. Q6NY19
Gene Alias 0610013D04Rik; Ankrd47; ankyrin repeat domain 47; ankyrin repeat domain-containing protein 47; D17Ertd288e; Kank3; kidney ankyrin repeat-containing protein 3; KN motif and ankyrin repeat domain-containing protein 3; KN motif and ankyrin repeat domains 3; NG28; putative KN motif and ankyrin repeat domains 3
Gene Symbols KANK3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KANK3. Recombinant protein control fragment (Product #RP-99982).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256949
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.