Learn More
Diagnostic Biosystems KAPPA LIGHT CHAIN Kappa Light Chain
Supplier:  Diagnostic Biosystems RP02501
Encompass_Preferred
This antibody reacts with free as well as bound kappa light chains. Contaminating antibodies have been removed by solid phase adsorption.
Catalog No. 50-238-5527
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.