Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kappa Light Chain Monoclonal Antibody (TB28-2), APC, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 17997042
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-997-042 100 Tests
17-997-041 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-997-042 Supplier Invitrogen™ Supplier No. 17997042
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: The monoclonal antibody TB28-2 recognizes the kappa light chain of human immunoglobulin. Approximately 22-kDa, the Ig kappa chain is comprised of one variable and one constant region. Two kappa chains together with the heavy chain comprise the whole Ig structure. Expression of Ig kappa is typically found on more than half of B lymphocytes. Applications Reported: This TB28-2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TB28-2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (1.0 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser

Immunoglobulin classes share the same basic four polypeptide chain structure of two heavy chains (five types) and two light chains (kappa, lambda; both having a molecular weight of 22.5 kDa). Kappa and lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region. The ratio of kappa to lambda is 70:30. Antibodies to the kappa light chain are reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kappa Light Chain
Applications Flow Cytometry
Classification Monoclonal
Clone TB28-2
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene IGK
Gene Accession No. P01601, P01602, P01834, P0DOX7
Gene Alias D4Mit8; HCAK1; ig kappa chain c region; Igk; Igk@; Igkc; Igk-C; IGKCD; IGKV; IGKV15; IGKV1-5; Immunoglobulin K; immunoglobulin kappa chain complex; immunoglobulin kappa constant; immunoglobulin kappa light chain; immunoglobulin kappa locus; immunoglobulin kappa variable 1-5; kappa; Kappa LC; Km; L12; L12a; LOC500183; similar to NGF-binding Ig light chain; V1
Gene Symbols IGK, IGKC, IGKV1-5, IGKV1D-16
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28299, 28901, 3514, 50802
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.