Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ Frontline 300 Chemical and Fire Protection Coverall with Hood Opening
SDP

Catalog No. 18025675 Shop All Kappler, Inc. Products
Change view
Click to view available options
Size:
Small/Medium, Medium
Large/X-Large, X-Large
2X-Large/3X-Large, 3X-Large
4X-Large
5X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-025-675 5X-Large
18-025-672 Large/X-Large, X-Large
18-025-671 Small/Medium, Medium
18-025-673 2X-Large/3X-Large, 3X-Large
18-025-674 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 18-025-675 Supplier Kappler™ Supplier No. F3H429 SI 5X
This item is not returnable. View return policy

Specifications

Size 5X-Large
Material Frontline™
Color Silver
Ankle Style Attached Boots, Boot Overflaps
Cuff Style Attached Gloves
Closure Type Double Storm Flap with Hook and Loop Seal
Seam Type Bound, Piped, Sealed
Resistance Flame, Chemicals
Back Style Expanded
Certifications/Compliance ASTM F1001
Description Front entry, expanded back, double-taped seams, 2 exhalation valves, attached sock boots with boot flap, Butyl gloves with specially attached barrier glove liner and knitted Dupont™ Kevlar™ aramid fiber overglove, Non-skid sole, cone Inserts
For Use With (Application) Ideal for petrochemical operations, line maintenance, tank cleaning, refueling situations
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.