Learn More
Kappler™ ProVent™ 10,000 Limited-Use Lab Coats
SDP
Catalog No. 17988491G Shop All Kappler, Inc. Products
Change view
Click to view available options
Color:
Blue
White
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
17-988-491G White 4X-Large
19-315346 Blue Medium
19-315348 Blue X-Large
19-315349 Blue 2X-Large
19-315350 Blue 3X-Large
19-315345 Blue Small
19-315347 Blue Large
17-988-491A White Small
17-988-491B White Medium
17-988-491C White Large
17-988-491D White X-Large
17-988-491F White 3X-Large
17-988-491E White 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Catalog No. 17-988-491G Supplier Kappler™ Supplier No. PPS235 WH 4X
This item is not returnable. View return policy
Comfortable fit in a tailored style
Patented microporous film composite fabric offers a wide range of barrier protection against blood, body fluids and viral threats.
  • High moisture vapor transmission rate provides cool, comfort
  • Ideal for biohazard cleanup
  • Six snaps for front closure
  • Serged-seam stitching, breast pocket, two hip pockets
  • Knit collar and cuffs
Compliance
Passes ASTM 1670 blood penetration and ASTM 1671 viral penetration tests.
Specifications
Product Type General Purpose Lab Coat
Size 4X-Large
Gender Unisex
Material ProVent Fabric
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
No. of Pockets 3
Model MS235WH
Seam Type Serged
Certifications/Compliance ASTM 1670, ASTM 1671
Weight (English) 18 lb.
Weight (Metric) 8.2 kg