Learn More
Description
Patented microporous film composite fabric offers a wide range of barrier protection against blood, body fluids and viral threats.
- High moisture vapor transmission rate provides cool, comfort
- Ideal for biohazard cleanup
- Six snaps for front closure
- Serged-seam stitching, breast pocket, two hip pockets
- Knit collar and cuffs
Compliance
Passes ASTM 1670 blood penetration and ASTM 1671 viral penetration tests.
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | Small |
| Gender | Unisex |
| Material | ProVent Fabric |
| Color | Blue |
| Closure Type | Snaps |
| Disposable | Single-use |
| Sterility | Non-sterile |
| No. of Pockets | 3 |
| Model | MS235BU |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
