Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ Provent 10000 Open-Back Gowns
SDP

Catalog No. 17988496E Shop All Kappler, Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-988-496E 2X-Large
17-988-496A Small
17-988-496B Medium
17-988-496C Large
17-988-496D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-988-496E Supplier Kappler™ Supplier No. PPN108BU2X
This item is not returnable. View return policy

Wrap-Around Gown, Hook and Loop Neck Closure, Two Waist Ties, Knit Cuffs, Non-Sterile

Specifications

For Use With (Application) General purpose
Gender Unisex
No. of Pockets None
Size 2X-Large
Product Type General Purpose Gown
Material Composite Fabric
Color Blue
Closure Type Hook and Loop
Disposable Single-use
Sterility Non-sterile
Cuff Style Knit
Model MN108BU2X
Seam Type Ultrasonic
Certifications/Compliance ASTM 1670, ASTM 1671
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.