Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ ProVent™ 7000 Wrap-Around Gowns, Hook and Loop Closure
SDP

Catalog No. 18025647 Shop All Kappler, Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-025-647 Medium
18-025-650 2X-Large
18-025-648 Large
18-025-649 X-Large
18-025-711 4X-Large
18-025-710 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 18-025-647 Supplier Kappler™ Supplier No. PVS101 WHMD
Add to Cart
Add to Cart
This item is not returnable. View return policy

Breathable, microporous fabric gowns offer general hazard protection for a variety of work environments from maintenance to general healthcare.

  • Wrap-around gown with hook and loop neck fastener
  • Two waist ties, knit cuffs, non-sterile
  • Serged seams
  • Quantity 30 per case
  • Protects against particle hazards and light splash situations
  • Passes the ASTM F1670 blood penetration test

Specifications

For Use With (Application) Maintenance, Spray Painting, General Healthcare
Product Type General Purpose Gown
Gender Unisex
No. of Pockets None
Size Medium
Material Microporous Coated Fabric
Color White
Closure Type Hook and Loop
Disposable Single-use
Sterility Non-sterile
Cuff Style Knit
Model LS101WHMD
Seam Type Serged
Sleeve Style Long Sleeve
Certifications/Compliance ASTM 1670
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.