Learn More
Description
- Wrap-around gown with hook and loop neck fastener
- Two waist ties, knit cuffs, non-sterile
- Serged seams
- Quantity 30 per case
- Protects against particle hazards and light splash situations
- Passes the ASTM F1670 blood penetration test
Specifications
Specifications
| Product Type | General Purpose Gown |
| For Use With (Application) | Maintenance, Spray Painting, General Healthcare |
| Gender | Unisex |
| No. of Pockets | None |
| Size | 3X-Large |
| Material | Microporous Coated Fabric |
| Color | White |
| Closure Type | Hook and Loop |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
