Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ ProVent™ 7000 Isolation Gowns, Tie Closures Neck and Waist
SDP

Catalog No. 19028300 Shop All Kappler, Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Cuff Style:
Elastic
Knit
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Cuff Style
19-028-300 Medium Knit
19-999-344D Small Elastic
19-999-344C Medium Elastic
19-999-344B Large Knit
19-028-298 Large Elastic
19-028-299 X-Large Elastic
19-999-344E X-Large Knit
19-999-344F 2X-Large Knit
19-028-301 2X-Large Elastic
18-025-653 3X-Large Elastic
19-999-344A Small Knit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 19-028-300 Supplier Kappler™ Supplier No. PVS112WHMD2H
Add to Cart
Add to Cart

Breathable, microporous fabric gowns offer general hazard protection for a variety of work environments from maintenance to general healthcare. Choose from elastic wrist or knit cuff options.

Lightweight composite fabric utilizes microporous film technology to provides protection against hazardous particles and light liquid splash and maintain strength wet or dry

  • Wraparound style opens to back
  • Knit cuffs or elastic wrists
  • Two neck ties, two waist ties
  • Nonsterile
  • White

Order Info

  • Front length: small 38.75 in. (98.4 cm), medium 39.75 in. (101 cm), large 40.75 in. (103.5 cm). Back length of all sizes: 3 in. (7.6 cm) longer than front

Compliance

Meets ASTM™ F1670 and ASTM F1671 standards.

Specifications

Gender Unisex
No. of Pockets None
For Use With (Application) Maintenance, Spray Painting, Healthcare, Clinical
Style Wrap-around
Product Type General Purpose Gown
Size Medium
Material Microporous Coated Fabric
Color White
Closure Type Neck and Waist Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Knit
Model PVS112WHMD2H
Seam Type Serged
Sleeve Style Long Sleeve
Certifications/Compliance ASTM F1670, ASTM F1671
Weight (English) 17 lbs.
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.