Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ Zytron™ 100 Chemical-Resistant Coveralls
SDP

Catalog No. 19167841F Shop All Kappler, Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Seam Type:
Bound
Serged
Includes:
Attached hood and boots, elastic wrists
Attached hood, elastic wrists and ankles
Collar, elastic wrists and ankles
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Seam Type Includes
19-167-841F 3X-Large Serged Attached hood and boots, elastic wrists
19-167-838C Large Bound Collar, elastic wrists and ankles
19-167-838B Medium Bound Collar, elastic wrists and ankles
19-167-839B Medium Serged Attached hood, elastic wrists and ankles
19-167-838E 2X-Large Bound Collar, elastic wrists and ankles
19-167-839C Large Serged Attached hood, elastic wrists and ankles
19-167-839E 2X-Large Serged Attached hood, elastic wrists and ankles
19-167-840C Large Bound Attached hood, elastic wrists and ankles
19-167-840B Medium Bound Attached hood, elastic wrists and ankles
19-167-840E 2X-Large Bound Attached hood, elastic wrists and ankles
19-167-841B Medium Serged Attached hood and boots, elastic wrists
19-167-841D X-Large Serged Attached hood and boots, elastic wrists
19-167-842D X-Large Bound Attached hood and boots, elastic wrists
19-167-842E 2X-Large Bound Attached hood and boots, elastic wrists
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 19-167-841F Supplier Kappler™ Supplier No. Z1S414XP2X3X
Add to Cart
Add to Cart
This item is not returnable. View return policy

Provides basic chemical resistance

  • Zippered front
  • Seam type available in bound and serged
  • Available with attached hood and/or boots and/or elastic wrists and ankles
  • Ideal for applications where workers need protection from dry particulates and light splash

Compliance

Meets ASTM™ F739 industrial chemical testing requirements.

Specifications

Size 3X-Large
Material Zytron™
Color Yellow
Ankle Style Elastic, Attached Boots
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper
Seam Type Serged
Resistance Chemicals
Certifications/Compliance ASTM D751, D3787
Description Attached hood and boots, elastic wrists
Includes Attached hood and boots, elastic wrists
For Use With (Application) Ideal for applications where workers need protection from dry particulates and light splash
Weight (English) 14 lb.
Weight (Metric) 6.4 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.