Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ Zytron™ 200 Coveralls
SDP

Catalog No. 19171136D Shop All Kappler, Inc. Products
Change view
Click to view available options
Description:
Elastic wrists and ankles
Plain wrists and ankles
Seam Type:
Bound
Sealed, Taped
Size:
Medium
Large
Large, X-Large
X-Large
2X-Large
2X-Large, 3X-Large
3X-Large
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Seam Type Size
19-171-136D Plain wrists and ankles Sealed, Taped 2X-Large
19-168-994 Elastic wrists and ankles Sealed, Taped Large, X-Large
19-168-995 Elastic wrists and ankles Sealed, Taped 2X-Large, 3X-Large
19-171-136A Plain wrists and ankles Sealed, Taped Medium
19-171-136B Plain wrists and ankles Sealed, Taped Large
19-171-136C Plain wrists and ankles Sealed, Taped X-Large
19-171-137A Elastic wrists and ankles Bound Medium
19-171-137B Elastic wrists and ankles Bound Large
19-171-137D Elastic wrists and ankles Bound 2X-Large
19-171-138A Elastic wrists and ankles Sealed, Taped Medium
19-171-138D Elastic wrists and ankles Sealed, Taped 2X-Large
19-171-135A Plain wrists and ankles Bound Medium
19-171-135B Plain wrists and ankles Bound Large
19-171-135C Plain wrists and ankles Bound X-Large
19-171-135D Plain wrists and ankles Bound 2X-Large
19-171-135E Plain wrists and ankles Bound 3X-Large
19-171-136E Plain wrists and ankles Sealed, Taped 3X-Large
19-171-137C Elastic wrists and ankles Bound X-Large
19-171-137E Elastic wrists and ankles Bound 3X-Large
19-171-138B Elastic wrists and ankles Sealed, Taped Large
19-171-138C Elastic wrists and ankles Sealed, Taped X-Large
19-171-138E Elastic wrists and ankles Sealed, Taped 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
This item is not returnable. View return policy
Catalog No. 19-171-136D Supplier Kappler™ Supplier No. Z2H4122X
This item is not returnable. View return policy

Resist wide range of chemicals

  • Suitable for uses where rip-out is a concern
  • Zippered front
  • Color: Gray

Compliance

Meets ASTM™ F739 industrial chemical testing requirements.

Specifications

Size 2X-Large
Material Zytron™
Color Gray
Ankle Style Open
Cuff Style Open
Neck Style Collared
Closure Type Front Zipper
Seam Type Sealed, Taped
Resistance Chemicals
Description Plain wrists and ankles
For Use With (Application) Ideal for wider range of chemicals and greater physical strength
Weight (English) 10 lb.
Weight (Metric) 4.5 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.