Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ Zytron™ 200 Hooded Coveralls
SDP

Catalog No. 19171142C Shop All Kappler, Inc. Products
Change view
Click to view available options
Description:
Attached hood, elastic wrist and face opening, attached sock boots with boot flap, double storm flap with hook and loop closure over the front zipper
Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening
Zipper front, attached hood and boots, elastic wrist and face opening, double storm flap with hook and loop closure
Zipper front, attached hood and boots, elastic wrist and face opening, double storm flap with hook and loop closure over the front zipper
Zipper front, attached hood and boots, elastic wrist and face opening, single storm flap with pressure sensitive tape closure
Seam Type:
Bound
Sealed, Taped
Size:
Medium
Small, Medium
Large
Large, X-Large
X-Large
2X-Large
2X-Large, 3X-Large
3X-Large
4X-Large
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Seam Type Size
19-171-142C Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening Bound X-Large
19-171-139B Zipper front, attached hood and boots, elastic wrist and face opening, single storm flap with pressure sensitive tape closure Bound Large
19-171-139A Zipper front, attached hood and boots, elastic wrist and face opening, single storm flap with pressure sensitive tape closure Bound Medium
19-171-139D Zipper front, attached hood and boots, elastic wrist and face opening, single storm flap with pressure sensitive tape closure Bound 2X-Large
19-171-140A Zipper front, attached hood and boots, elastic wrist and face opening, double storm flap with hook and loop closure Sealed, Taped Medium
19-171-140B Zipper front, attached hood and boots, elastic wrist and face opening, double storm flap with hook and loop closure Sealed, Taped Large
19-171-140D Zipper front, attached hood and boots, elastic wrist and face opening, double storm flap with hook and loop closure Sealed, Taped 2X-Large
19-171-141C Attached hood, elastic wrist and face opening, attached sock boots with boot flap, double storm flap with hook and loop closure over the front zipper Sealed, Taped Large, X-Large
19-171-141A Attached hood, elastic wrist and face opening, attached sock boots with boot flap, double storm flap with hook and loop closure over the front zipper Sealed, Taped Small, Medium
19-171-141E Attached hood, elastic wrist and face opening, attached sock boots with boot flap, double storm flap with hook and loop closure over the front zipper Sealed, Taped 2X-Large, 3X-Large
19-171-142B Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening Bound Large
19-171-142A Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening Bound Medium
19-171-143D Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening Sealed, Taped 2X-Large
19-171-142D Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening Bound 2X-Large
19-171-143B Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening Sealed, Taped Large
19-171-143A Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening Sealed, Taped Medium
19-167-811A Zipper front, attached hood and boots, elastic wrist and face opening, double storm flap with hook and loop closure over the front zipper Sealed, Taped 4X-Large
19-171-139C Zipper front, attached hood and boots, elastic wrist and face opening, single storm flap with pressure sensitive tape closure Bound X-Large
19-171-139E Zipper front, attached hood and boots, elastic wrist and face opening, single storm flap with pressure sensitive tape closure Bound 3X-Large
19-171-142E Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening Bound 3X-Large
19-171-143C Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening Sealed, Taped X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
This item is not returnable. View return policy
Catalog No. 19-171-142C Supplier Kappler™ Supplier No. Z2B428XL
This item is not returnable. View return policy

Provide increased protection with resistance to a wider range of chemicals

  • Attached hood
  • Zippered front
  • Color: Gray

Applications:

Valuable in applications where rip-out is a concern

Specifications

Size X-Large
Material Zytron™
Color Gray
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Front Zipper
Seam Type Bound
Resistance Chemicals
Certifications/Compliance ASTM D751, D3787
Description Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening
For Use With (Application) Ideally used with wider range of chemicals
Weight (English) 16 lb.
Weight (Metric) 7.3 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.