Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ Zytron 200 Totally Encapsulating Suits
SDP

Catalog No. 19171145C Shop All Kappler, Inc. Products
Change view
Click to view available options
Description:
Expanded back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper
Expanded back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper
Flat back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper
Flat back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper
Seam Type:
Bound, Taped
Sealed, Taped
Size:
Medium
Large
X-Large
2X-Large
3X-Large
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Seam Type Size
19-171-145C Flat back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped X-Large
19-171-145A Flat back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped Medium
19-171-145B Flat back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped Large
19-171-145D Flat back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped 2X-Large
19-171-146B Expanded back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped Large
19-171-146D Expanded back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped 2X-Large
19-171-146A Expanded back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped Medium
19-171-147A Expanded back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped Medium
19-171-147B Expanded back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped Large
19-171-147D Expanded back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped 2X-Large
19-171-144A Flat back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped Medium
19-171-144B Flat back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped Large
19-171-144D Flat back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped 2X-Large
19-171-144C Flat back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped X-Large
19-171-144E Flat back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped 3X-Large
19-171-145E Flat back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped 3X-Large
19-171-146C Expanded back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped X-Large
19-171-146E Expanded back, rear entry, side air inlet, single storm flap with pressure sensitive tape closure over the front zipper Bound, Taped 3X-Large
19-171-147C Expanded back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped X-Large
19-171-147E Expanded back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper Sealed, Taped 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
This item is not returnable. View return policy
Catalog No. 19-171-145C Supplier Kappler™ Supplier No. Z2H371XL
This item is not returnable. View return policy

Specifications

Size X-Large
Material Zytron™
Color Gray
Closure Type Double Storm Flap with Hook and Loop Seal
Seam Type Sealed, Taped
Resistance Chemicals
Back Style Flat
Description Flat back, rear entry, side air inlet, double storm flap with hook and loop closure, cloth zipper
For Use With (Application) Ideally used with wider range of chemicals
Weight (English) 12 lb.
Weight (Metric) 5.4 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.