Learn More
Selleck Chemical LLC Kartogenin 10mg 4727-31-5
SDP
Supplier:  Selleck Chemical LLC S765810MG
Encompass_Preferred
Kartogenin is an activator of the smad4/smad5 pathway, and promotes the selective differentiation of multipotent mesenchymal stem cells into chondrocytes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5088
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.