Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam KAT13D / CLOCK Synthetic Peptide, Unconjugated, >95% Purity, 50 ug

Catalog No. 502710376
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-0376 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-0376 Supplier Abcam Supplier No. AB585450UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic KAT13D / CLOCK peptide with >95% purity for blocking applications.

  • Target Protein: KAT13D / CLOCK
  • Purity: >95%
  • Applications: Suitable for blocking (BL)
  • Type: Synthetic peptide

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Concentration 0.35 mg/mL
Content And Storage Store at -20°C.
Form Solution
Label or Dye Unconjugated
Substrate Properties Peptide-based Substrate
Target Molecule KAT13D, CLOCK
Quantity 50 μg
For Use With (Application) Blocking, Neutralization
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.