Learn More
Medchemexpress LLC HY-N2096 500mg Medchemexpress, Kavain CAS:3155-48-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN2096500MG
Encompass_Preferred
Medchemexpress, HY-N2096 500mg Kavain CAS:3155-48-4 Kavain is a class of kavalactone isolated from Piper methysticum, which has anxiolytic and sedative properties in animals and humans. Kavain positively modulated γ-Aminobutyric acid type A (GABAA) receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2862
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.