Learn More
Kawasumi™ Gravity IV Administration Sets: Y-Needle
Catalog No. 22010102
Change view
Click to view available options
Length (English) Tube:
84 in.
100 in.
Product Type:
Dual Y-needle Injection Set
Y-needle IV Adminstration Set
Quantity:
10 drops/mL
60 drops/mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Tube Product Type Quantity
22-010-102 100 in. Dual Y-needle Injection Set 10 drops/mL
22-010-104 84 in. Y-needle IV Adminstration Set 10 drops/mL
22-010-106 84 in. Y-needle IV Adminstration Set 60 drops/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-010-102 Supplier Kawasumi™ Supplier No. IV016Y
Add to Cart
Add to Cart
This item is not returnable. View return policy
A wide selection of sets and components meet a variety of solution administration requirements
  • Low cost gravity IV administration set
  • Sets feature Non-DEHP fluid paths, including tubing and drip chamber
  • Y-needle style
  • Latex-free
Specifications
Product Type Dual Y-needle Injection Set
For Use With (Application) Gravity IV Administration
Length (English) Tube 100 in.
Length (Metric) Tube 254 cm
Quantity 10 drops/mL