Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KBTBD8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584805
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584805 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584805 Supplier Invitrogen™ Supplier No. PA584805
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NKWTRKKDFP CDQSINPYLK LVLFQNKLHL FVRATQVTVE EHVFRTSRKN SLYQYDDIAD QWMKVYETPD RLWDLGRHFE CAVAKLYPQC LQK Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.

KBTBD8 (Kelch Repeat And BTB Domain Containing 8) is a Protein Coding gene. Diseases associated with KBTBD8 include Treacher Collins Syndrome 1. Among its related pathways are Class I MHC mediated antigen processing and presentation and Innate Immune System. An important paralog of this gene is KBTBD2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KBTBD8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KBTBD8
Gene Accession No. Q8NFY9
Gene Alias AU040152; cb672; EMBL:AAI60902.1, ECO:0000312; kbtbd8; kbtbd8 {ECO:0000312; kelch repeat and BTB (POZ) domain containing 8; kelch repeat and BTB domain containing 8; kelch repeat and BTB domain-containing protein 8; kelch repeat and BTB domain-containing protein 8 {ECO:0000250; KIAA1842; LOW QUALITY PROTEIN: kelch repeat and BTB domain-containing protein 8; mKIAA1842; RGD:1563166}; RGD1563166; SSEC 51; SSEC51; SSEC-51; TAKRP; TA-KRP; T-cell activation kelch repeat protein; UniProtKB:Q8NFY9}; wu:fc57a10; zgc:152950
Gene Symbols KBTBD8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KBTBD8. Recombinant protein control fragment (Product #RP-105465).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84541
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.